Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPI Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14315

Catalog No. PA14315


Only null left
Add to Cart

Description

Description

PA1-4315 detects the SHPRH protein in mouse samples. PA1-4315 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~200 kDa protein representing SHPRH in mouse brain lysate. PA1-4315 immunizing peptide corresponds to amino acid residues 515-535 of the mouse SHPRH protein. There is 95% sequence identity to rat, and 90% sequence identity to human.

A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CENPI
Polyclonal
Unconjugated
Cenpi
2610103K11Rik; AA450458; AU024614; BC006883; CENPI; CENP-I; Centromere protein I; D230017O13Rik; E130018M05; follicle-stimulating hormone primary response protein; FSH primary response 1; FSH primary response protein 1; FSHPRH1; ICEN19; Interphase centromere complex protein 19; leucine-rich primary response protein 1; LRPR1; Mis6
Rabbit
Antigen Affinity Chromatography
RUO
102920
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8K1K4
Cenpi
Synthetic peptide: D(515) K T K K Q A V G S P R K I E K E L R K S(535).
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.