Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPN Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179664
Description
CENPN Polyclonal specifically detects CENPN in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CENPN | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
NP_001094095 | |
CENPN | |
Synthetic peptide directed towards the middle region of human CENPNThe immunogen for this antibody is CENPN. Peptide sequence SFKKILQRALKNVTVSFRETEENAVWIRIAWGTQYTKPNQYKPTYVVYYS. | |
Affinity Purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Dog: 86%; Pig: 86%; Bovine: 86%; Guinea pig: 86%; Rat: 79%; Mouse: 79%. | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
BM039, C16orf60, CENP-N, centromere protein N, chromosome 16 open reading frame 60, FLJ13607, FLJ22660, ICEN32, Interphase centromere complex protein 32 | |
Rabbit | |
41 kDa | |
100 μL | |
Stem Cell Markers | |
55839 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title