Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPP Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen CENPP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP153064
View Documents
Novus Biologicals
NBP153064
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

CENPP Polyclonal specifically detects CENPP in Human samples. It is validated for Western Blot.
Specifications

Specifications

CENPP
Polyclonal
Rabbit
Cell Cycle and Replication
Q6IPU0
401541
Synthetic peptides corresponding to CENPP(centromere protein P) The peptide sequence was selected from the N terminal of CENPP. Peptide sequence VQKSFQAIHQFNLEGWKSSKDLKNQLGHLESELSFLSTLTGINIRNHSKQ.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
PBS and 2% Sucrose with 0.09% Sodium Azide
CENP-PRP11-19J3.3, centromere protein P, FLJ33928
CENPP
IgG
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit