Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPQ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544696
Description
Peptide sequence: VRNTVKKNKN HLKDLSSEGQ TKHTNLKHGK TAASKRKTWQ PLSKSTRDHL Sequence homology: Human: 100%.
CENPQ is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
Specifications
CENPQ | |
Polyclonal | |
Unconjugated | |
CENPQ | |
C6orf139; CENPQ; CENP-Q; Centromere protein Q | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55166 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7L2Z9 | |
CENPQ | |
Synthetic peptide directed towards the N-terminal of human CENPQ (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction