Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPU Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544981
Description
Peptide sequence: ISHDKRKKSR SKAIGSDTSD IVHIWCPEGM KTSDIKELNI VLPEFEKTHL Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
CENPU | |
Polyclonal | |
Unconjugated | |
Cenpu | |
1700029A22Rik; CENP50; CENP-50; Cenpu; CENP-U; CENPU50; Centromere protein of 50 kDa; centromere protein U; ICEN24; Interphase centromere complex protein 24; KLIP1; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; LOC617866 protein; MLF1 interacting protein; MLF1-interacting protein; MLF1IP; myeloid leukemia factor 1 interacting protein; PBIP1; Polo-box-interacting protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79682 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q71F23 | |
Cenpu | |
Synthetic peptide directed towards the C-terminal of human MLF1IP (aa 214-263). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction