Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPU Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550454
Description
The antibody detects endogenous levels of total CENPU protein.
CENPU Polyclonal antibody specifically detects CENPU in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CENPU | |
Polyclonal | |
Unconjugated | |
Cenpu | |
1700029A22Rik; CENP50; CENP-50; Cenpu; CENP-U; CENPU50; Centromere protein of 50 kDa; centromere protein U; ICEN24; Interphase centromere complex protein 24; KLIP1; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; LOC617866 protein; MLF1 interacting protein; MLF1-interacting protein; MLF1IP; myeloid leukemia factor 1 interacting protein; PBIP1; Polo-box-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79682 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q71F23 | |
Cenpu | |
Full length fusion protein of Human CENPU | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction