Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Centaurin alpha-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528723
Description
Recommended positive controls: ADAP2-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GTPase-activating protein for the ADP ribosylation factor family (Potential). Binds phosphatidylinositol 3, 4, 5-trisphosphate (PtdInsP3) and inositol 1, 3, 4, 5-tetrakisphosphate (InsP4). Possesses a stoichiometry of two binding sites for InsP4 with identical affinity.
Specifications
Centaurin alpha-2 | |
Polyclonal | |
Unconjugated | |
ADAP2 | |
Adap2; Arf-GAP with dual PH domain-containing protein 2; ArfGAP with dual PH domains 2; CENTA2; centaurin; centaurin beta; centaurin, alpha 2; centaurin-alpha 2 protein; centaurin-alpha-2; Centaurin-alpha2 protein; cent-b; cnt-a2; HSA272195 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55803 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.49 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9NPF8 | |
ADAP2 | |
Synthetic peptide corresponding to a region within amino acids 319 and 381 of Human centaurin-alpha 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction