Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550542

Catalog No. PA550542


Add to cart

Description

Description

The antibody detects endogenous levels of total Centaurin gamma 2 protein.

CENTG2 belongs to an ADP-ribosylation factor GTPase-activating (ARF-GAP) protein family involved in membrane traffic and actin cytoskeleton dynamics (Nie et al., 2002 [PubMed 12388557]).
Specifications

Specifications

AGAP1
Polyclonal
Unconjugated
AGAP1
Liquid
AGAP1
Synthetic peptide corresponding to a region derived from internal residues of human ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9UPQ3, Q8BXK8
CENTG2, KIAA1099
Rabbit
IgG
100 μL
-20°C
Primary
116987, 347722
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit