CENTiCARE Deluxe Phlebotomy Carts with 3 in. Casters
Multi-purpose use carts for phlebotomy or nurse collection cart needs with tray in nursing units
Manufacturer: CENTiCARE CPHL1401H
 View more versions of this product
Catalog No. 22-555-100
  Check Availability
Add to Cart
    Specifications
    No. of Casters 5
    Product Type Deluxe phlebotomy cart
    Design Modular
    Wheel Type Silent deluxe casters
    For Use With (Application) Phlebotomy or nurse collection needs with tray on nursing units
    For Use With (Equipment) Fisher cat. 22-267-270 and 22-267-271 blood collection tray (not included)
    Includes
    • Writing surface
    • Tray holder
    • Handle
    • 16.5 x 10.8 x 3.5 in. clear bin with bracket and three dividers
    • 5 quart Sharps bracket
    Material Stainless steel
    Warranty 180 days limited liability
  • Description & Specifications
Related Products
Product Content Feedback