Learn More
Applied Biosystems™ Centri-Sep™ 96-Well Plates
Centri-Sep™ 96's hydrated matrix enables effective and reliable high-throughput removal of excess BigDye™ terminators from DNA sequencing reactions, giving you clean sequencing reads with uniform signal intensity starting with the first base.
$4385.00
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
43-678-21 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
4367821
N/A
Each for $4,385.00
Add to cart
43-678-19 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
4367819
N/A This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Processed faster than ultrafiltration columns
  • Rapid and efficient separations
  • Buffer is not preselected
  • Convenient 20 to 100μL sample size
  • Dry gel format
  • Room Temperature Storage
  • Microcentrifuge format