Learn More
Daigger & CompanySupplier Diversity Partner CENTRIFUGAL INSERT 3K PK12 DFS Item
Manufacturer:  Daigger & CompanySupplier Diversity Partner OD003C65
Pall Life Sciences Jumbosep Centrifugal Devices Membrane Insert Only 3K Gray Concentrates 60mL Volumes to 5mL in less than 30 Minutes PK/12
Catalog No. 50-112-9789