Learn More
Chemglass Life Sciences Mini Centrifuge, 4,000rpm
SDP
Supplier:  Chemglass Life Sciences CLS1601002
Encompass_Preferred
Mini Centrifuge, 4,000rpm. • Quiet, vibration-free operation• Include two, easy-to-change rotors• One-year warrantyThese mini centrifuges are popular for quick spins, microfiltration of samples, and cell separations. The clear covers are made of high strength plastic and provide easy viewing of samples. These centrifuges quickly accelerate to their maximum speed and stop within seconds. Manufactured to precise specifications, resulting in smooth runs that are quiet and vibration-free. Included is a round rotor with capacity for six standard microtubes (1.5 to 2.0mL) as well as a strip tube rotor for 2 x 8 PCR strips or sixteen individual 0.2mL tubes. The rotors can be easily interchanged by the user within a matter of seconds. Other RPMs are available, please contact us for details.
Catalog No. 50-143-459