Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Centrin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580470
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CETN2/Centrin 2.
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Specifications
Centrin 2 | |
Polyclonal | |
Unconjugated | |
CETN2 | |
1110034A02Rik; AI326150; CALT; Caltractin; caltractin (20kD calcium-binding protein); Caltractin isoform 1; caltractin, 20 kDa calcium binding protein; CEN2; centrin 2; centrin, EF-hand protein, 2; Centrin2; centrin-2; Cetn2; EF hand protein | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
1069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
P41208 | |
CETN2 | |
Recombinant Human CETN2/Centrin 2 Protein (Met1-Tyr171). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction