Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centrin 3 Antibody (OTI4G5) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270409

Catalog No. NBP270409


Add to Cart

Description

Description

Centrin 3 Monoclonal specifically detects Centrin 3 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Centrin 3
Monoclonal
LYOPH
Western Blot 1:2000
CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245
Mouse
19.4 kDa
100 μg
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Western Blot
OTI4G5
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
CETN3
Full length human recombinant protein of human CETN3 (NP_004356) produced in E.coli.
Affinity Purified
Cell Cycle and Replication
1070
Human, Mouse
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit