Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centrin 3 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270409AF647

Catalog No. N270409A647


Add to cart

Description

Description

Centrin 3 Monoclonal antibody specifically detects Centrin 3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Centrin 3
OTI4G5
Western Blot
Affinity Purified, Conjugated
CETN3
Full length recombinant protein of human Centrin 3 (NP_004356) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
1070.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit