Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin 3 Mouse anti-Human, DyLight 755, Clone: OTI4G5, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270409IR
Description
Centrin 3 Monoclonal antibody specifically detects Centrin 3 in Human samples. It is validated for Western Blot.Specifications
Centrin 3 | |
OTI4G5 | |
Western Blot | |
Affinity Purified, Conjugated | |
CETN3 | |
Full length recombinant protein of human Centrin 3 (NP_004356) produced in E.coli. | |
Immunogen affinity purified | |
Cell Cycle and Replication | |
Primary | |
1070.0 |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title