Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP First Responder Spill Kits


Click to view available options
Size:
2 Responders
4 Responders
6 Responders
8 Responders
Description
Includes all necessary supplies to safely contain and clean up oils, acids, caustics, solvents, and toxic spills; also includes protective apparel for the number of persons indicated in the kit size (2, 4, 6 or 8 responders).
- One 95-gal. poly overpack drum
- 100 17 × 19 in. universal pads
- Four 5 × 10 in. universal booms
- Four epoxy sticks
- One pipe wrap kit
- Two hazardous waste labels
- One nonsparking shovel
- One 36 × 36 in. polyurethane drain seal
- One quart each of acid and caustic neutralizer
- Coveralls with hood and booties, nitrile gloves, safety goggles, and disposal bags to accommodate number of persons designated in kit size
Specifications
Specifications
Capacity (English) | 95 gal. |
Capacity (Metric) | 359.6 L |
For Use With (Application) | Clean up oils, acids, caustics, solvents, and toxic spills |
Includes | 95 gal. poly overpack drum, (4) 5 x 10 in. universal booms, 4 epoxy sticks, 1 pipe wrap kit, 2 hazardous waste labels, 1 non-sparking shovel, 36 x 36 in. polyurethane drain seal, 1 quart each of acid and caustic neutralizer |
Size | 4 Responders |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction