Learn More
CEP™ Oil-Only Blue Fine-Fiber Bonded Sorbents
Absorbent and perforated, allowing precise measurement
Manufacturer:  COMPLETE ENVIRONMENTAL PRODUCTS QBEP100
 View more versions of this product
Catalog No. 19-130-5845
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Blue
113.5L
17 in.
For heavy spills, absorbent and perforated, allowing precise measurement
Medium
15 in.
30 gal., 113.5L
30 gal.
43.18cm
Universal Fine Bonded Sorbent Pad
Polypropylene Fiber
Medium
38cm
17 x 15 in. (43.18 x 38cm)