Learn More
CEP Oil Only Booms, Pillows and Socks
Variety of sorbents for all types of oil spills
Manufacturer:  CEP B410
 View more versions of this product
Catalog No. 19-056-200
Add to cart
Description
Description
  • Hydrophobic, hydrophilic, static resistant
  • Nonlinting or anti-shed netting
  • Overlapping clips for deployment in any length
  • Pillows and socks are ideal for catch basins and sumps
Specifications
Specifications
Safety
4 in.
3.04m
Sorbent Boom
White
10.1cm
46 in.
For oil spill control, skimming oil off water and for oil spill clean-up