Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP Oil-Only Fine-Fiber Bonded Sorbents

Exceptionally strong oil-only and universal sorbents

$89.40 - $467.50

Specifications

Color White
For Use With (Application) For heavy spills, keep floors clean, dry, and safe
Material Polypropylene Fiber
View More Specs

Products 7
Catalog Number Mfr. No. Length (English) Weight Capacity Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Weight Capacity Width (English) Price Quantity & Availability  
19-150-0238
View Documents
CEP
Q24
150 ft. Heavy 24 in.
Each for $455.50
Add to cart
 
19-150-0235
View Documents
CEP
Q200
17 in. Light 15 in.
Case of 200 for $89.40
Add to cart
 
19-150-0239
View Documents
CEP
Q288
300 ft. Light 30 in.
Each for $435.00
Add to cart
 
19-150-0241
View Documents
CEP
Q288S
300 ft. Heavy 30 in.
Case of 2 for $435.00
Add to cart
 
19-150-0236
View Documents
CEP
Q50
30 in. Heavy 30 in.
Case of 50 for $467.50
Add to cart
 
19-150-0234
View Documents
CEP
QEP100
17 in. Medium 15 in.
Case of 100 for $217.50
Add to cart
 
19-150-0240
View Documents
CEP
QSR150
150 ft. Heavy 15 in.
Case of 2 for $442.00
Add to cart
 
Description

Description

  • Constructed of two layers of low-linting, fine-fiber polypropylene and ultrasonically bonded around a high-loft meltblown core
  • Dimpled for faster adsorption—ideal for heavy spills
  • Perforated to extend use before disposal
  • Control oversprays and spills with excellent absorbency

Specifications

Specifications

White
For heavy spills, keep floors clean, dry, and safe
Polypropylene Fiber
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit