Learn More
CEP Oil-Only Fine-Fiber Bonded Sorbents
Exceptionally strong oil-only and universal sorbents
Manufacturer:  COMPLETE ENVIRONMENTAL PRODUCTS Q288S
 View more versions of this product
Catalog No. 19-150-0241
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
White
204.4L
300 ft.
For heavy spills, keep floors clean, dry, and safe
Heavy
76.2cm
300 ft. x 30 in. (91.4m x 76.2cm)
54 gal.
91.4m
Bonded Sorbent Split Roll
Polypropylene Fiber
30 in.
54 gal., 204.4L