Learn More
CEP Oil Only Pads and Rolls
Contain and adsorb oils while repelling aquatic liquids
Manufacturer:  CEP CEPR144
 View more versions of this product
Catalog No. 19-057-832
Add to cart
Description
Description
  • Constructed of meltblown polypropylene fibers for cost-effective adsorption
  • Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
  • Choice of pads and rolls with varying sorbing capacities
Specifications
Specifications
Safety
189L
150 ft.
Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
Heavy
76.2cm
50 gal.
45.7m
Sorbent Roll
Polypropylene Fiber
30 in.
150 ft. x 30 in. (45.7m x 76.2cm)