Learn More
CEP Oil Only Pads and Rolls
Contain and adsorb oils while repelling aquatic liquids
Manufacturer:  CEP CEPEP50
 View more versions of this product
Catalog No. 19-500-528
Add to cart
Description
Description
  • Constructed of meltblown polypropylene fibers for cost-effective adsorption
  • Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
  • Choice of pads and rolls with varying sorbing capacities
Specifications
Specifications
Safety
131.7L
30 in.
Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
Medium
76.2cm
34.8 gal.
76.2cm
Sorbent Pad
Polypropylene Fiber
30 in.
30 x 30 in. (76.2 x 76.2cm)