Learn More
CEP Oil Only Pads and Rolls
Contain and adsorb oils while repelling aquatic liquids
$26.48 - $399.00
Specifications
For Use With (Application) | Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon |
---|---|
Material | Polypropylene Fiber |
Catalog Number | Mfr. No. | Capacity (English) | Length (English) | Weight Capacity | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Length (English) | Weight Capacity | Width (English) | Price | Quantity & Availability | ||
19-500-528
|
CEP
CEPEP50 |
34.8 gal. | 30 in. | Medium | 30 in. |
Case of 50 for $262.50
|
|
19-057-832
|
CEP
CEPR144 |
50 gal. | 150 ft. | Heavy | 30 in. |
Each for $399.00
|
|
19-057-831
|
CEP
CEPP100 |
24 gal. | 17 in. | Heavy | 15 in. |
Pack of 100 for $231.00
|
|
19-500-530
|
CEP
CEPP50 |
44 gal. | 30 in. | Heavy | 30 in. |
Case of 50 for $320.50
|
|
19-500-531
|
CEP
CEPR288 |
52 gal. | 300 ft. | Single | 15 in. |
Each for $348.00
|
|
19-150-0150
|
CEP
L100 |
13.5 gal. | 17 in. | Single | 15 in. |
Case of 100 for $26.48
|
|
19-051-755
|
CEP
CEPEP100 |
19 gal. | 17 in. | Medium | 15 in. |
Case of 100 for $26.65
|
|
19-500-529
|
CEP
CEPESR144 |
39 gal. | 150 ft. | Medium | 15 in. |
Case of 2 for $239.00
|
|
19-500-532
|
CEP
CEPSR144 |
50 gal. | 150 ft. | Heavy | 15 in. |
Case of 2 for $242.50
|
|
19-057-830
|
CEP
CEPP200 |
24 gal. | 17 in. | Single | 15 in. |
Case of 200 for $238.00
|
|
19-811-285
|
CEP
P200 |
25.2 gal. | 17 in. | Single | 15 in. |
Pack of 200 for $236.50
|
|
Description
- Constructed of meltblown polypropylene fibers for cost-effective adsorption
- Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
- Choice of pads and rolls with varying sorbing capacities
Specifications
Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon | |
Polypropylene Fiber |