Specifications
EPA | |
Attaches to a Poly-Stacker and allows simultaneous dispensing from all drums | |
1.89 L | |
Poly-Shelf™ | |
2400 lb. | |
17 x 22 x 17.5 in. (43.18 x 55.88 x 44.45 cm) | |
43.18 cm | |
55.88 cm | |
44.45 cm |
Yellow | |
0.5 gal. | |
Polyethylene | |
Manufacturing plants, mining, chemical storage facilities | |
1088.6 kg | |
17 in. | |
22 in. | |
17.5 in. |