Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP Universal Aggresive Booms, Pillows, and Socks
SDP

Ideal for containing and absorbing spills of any liquid

$367.50 - $372.00

Specifications

For Use With (Application) Containing and absorbing spills of any liquid
View More Specs

Products 2
Catalog Number Mfr. No. Color Capacity (English) Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Capacity (English) Length (English) Price Quantity & Availability  
19-135-955
SDP
View Documents
CEP
HAZSOC12
Orange 20 gal. 46 in.
Case of 12 for $372.00
Add to Cart
 
19-135-956
SDP
View Documents
CEP
HAZSOC10
Orange 18 gal. 10 ft.
Case of 4 for $367.50
Add to Cart
 
Description

Description

  • All products absorb up to 25 times their weight in fluid
  • Color coded: universal booms are striped; socks are orange; and pillows are white

Specifications

Specifications

Containing and absorbing spills of any liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit