Learn More
CEP Universal Aggresive Booms, Pillows, and Socks
Ideal for containing and absorbing spills of any liquid
Manufacturer:  COMPLETE ENVIRONMENTAL PRODUCTS HAZPIL10
 View more versions of this product
Catalog No. 19-135-957
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
White
151.4L
24 in.
For containing and absorbing spills of any liquid
7.62cm
45.72cm
40 gal.
60.9cm
Universal Hazmat Sorbent Pillow
3 in.
18 in.
24 x 18 x 3 in. (60.9 x 45.72 x 7.62cm)