Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP152 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550004

Catalog No. PA550004


Add to cart

Description

Description

The antibody detects endogenous levels of total CEP152 protein.

Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
Specifications

Specifications

CEP152
Polyclonal
Unconjugated
CEP152
Liquid
CEP152
Synthesized peptide derived from internal of human CEP152.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O94986, A2AUM9
KIAA0912
Rabbit
IgG
100 μL
-20°C
Primary
22995, 99100
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit