Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP164 Mouse anti-Human, DyLight 650, Clone: 13, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243631C

Catalog No. NBP243631C


Add to cart

Description

Description

CEP164 Monoclonal antibody specifically detects CEP164 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CEP164
13
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
CEP164
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
centrosomal protein 164kDa, FLJ54767
Mouse
IgG1
0.1 ml
Primary
22897
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit