Learn More
CEP170/CEP170L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188991AP
Catalog No. 50-173-2276
Add to cart
Description
Description
This locus appears to be a transcribed pseudogene similar to centrosomal protein 170 kDa (CEP170). An approximately 50 kb region upstream of this locus also is homologous to CEP170, but is not transcribed.
Specifications
Specifications
CEP170/CEP170L
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6A065, Q96L14
CEP170, CEP170L, FAM68B, KIAA0470, KIAA0470L
Rabbit
IgG
150 μL
-20°C
Primary
171457, 545389, 645455
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CEP170P1
Liquid
Cep170, CEP170P1
CEP170/CEP170L Fusion Protein Ag13532
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat