Learn More
CEP44 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 244571AP
Catalog No. 50-173-4370
Add to cart
Description
Description
This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer ft.s Disease.
Specifications
Specifications
CEP44
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5HZK1, Q9C0F1
CEP44, KIAA1712, PS1TP3
Rabbit
IgG
150 μL
-20°C
Primary
382010, 80817
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CEP44
Liquid
Cep44
CEP44 Fusion Protein Ag19961
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse