Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cep55 Mouse anti-Human, Clone: 5A3, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen Cep55
Clone 5A3
Concentration 1.0 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS2153100
View Documents
MilliporeSigma
MABS2153100UG
100 μg
Each for $373.00
Add to cart
 
MABS215325
View Documents
MilliporeSigma
MABS215325UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

Cep55 Monoclonal antibody specifically detects Cep55 in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Cep55
1.0 mg/mL
Unconjugated
Mouse
Human
Full-length recombinant human Centrosomal protein of 55 kDa (Cep55).
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
5A3
Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit