Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP55 Rabbit anti-Human, Mouse, Rat, Clone: S05-6A6, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | CEP55 |
---|---|
Clone | S05-6A6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CEP55 Monoclonal antibody specifically detects CEP55 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CEP55 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
C10orf3, cancer/testis antigen 111, centrosomal protein 55kDa, centrosomal protein of 55 kDa, Cep55, chromosome 10 open reading frame 3, CT111, FLJ10540, Up-regulated in colon cancer 6, URCC6 | |
A synthetic peptide of human CEP55 (Uniprot#: Q53EZ4) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S05-6A6 | |
Monoclonal | |
Purified | |
RUO | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
55165 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title