Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530551
Description
Recommended positive controls: NT2D1, MCF-7. Predicted reactivity: Mouse (94%), Rat (92%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).
Specifications
CEP57 | |
Polyclonal | |
Unconjugated | |
CEP57 | |
3110002L15Rik; 4921510P06Rik; 4931428M20Rik; AI467480; centrosomal protein 57; centrosomal protein 57kDa; centrosomal protein of 57 kDa; Cep57; FGF2-interacting protein; KIAA0092; mKIAA0092; MVA2; PIG8; proliferation-inducing protein 8; RGD1309884; testis-specific protein 57; Translokin; Tsp57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9702 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.94 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q86XR8 | |
CEP57 | |
Recombinant fragment corresponding to a region within amino acids 36 and 281 of Human CEP57. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction