Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP68 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI5B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271728JF646

Catalog No. N271728J646


Add to cart

Description

Description

CEP68 Monoclonal antibody specifically detects CEP68 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

CEP68
OTI5B4
Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
CEP68
Full length recombinant protein of human CEP68 (NP_055962) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
centrosomal protein 68kDa, centrosomal protein of 68 kDa, Cep68, FLJ36750, KIAA0582FLJ25920
Mouse
IgG1
0.1 mL
Primary
23177.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit