Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cephalexin, 50mg/mL in 1M ammonium hydroxide, sterile-filtered

Click to view available options
Quantity:
25mL
50mL
15686-71-2
C16H20ClN3O5S
401.862
MFCD00167148
YHJDZIQOCSDIQU-OEDJVVDHSA-N
57515946
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate
[H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-]
15686-71-2
C16H20ClN3O5S
401.862
MFCD00167148
YHJDZIQOCSDIQU-OEDJVVDHSA-N
57515946
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate
[H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-]
Physical Form Liquid
Quantity 50mL
Chemical Name or Material Cephalexin, 50mg/mL in 1M ammonium hydroxide, sterile-filtered

GHS H Statement
H334-H318-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P305+P351+P338-P310-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0350000

storageNote1 : -30° to -10°C