Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cephalothin sodium salt, 50mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
58-71-9
C16H15N2NaO6S2
418.414
MFCD00072025
VUFGUVLLDPOSBC-XRZFDKQNSA-M
23675321
CHEBI:3542
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+]
58-71-9
C16H15N2NaO6S2
418.414
MFCD00072025
VUFGUVLLDPOSBC-XRZFDKQNSA-M
23675321
CHEBI:3542
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+]
Physical Form Liquid
Quantity 1mL
Chemical Name or Material Cephalothin sodium salt, 50mg/mL in distilled water, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0388300

storageNote1 : -30° to -10°C