Cepheid™ SmartCycler™ Reaction Tubes

For use with the SmartCycler Real-Time Thermal Cycler

Manufacturer: Cepheid 9000085

 View more versions of this product

Catalog No. 9000085

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 25.0μL
    Volume (Metric) 25μL
    Quantity 250 Bag
    Cap Type With
    Format Tube
    For Use With (Equipment) SmartCycler™ real-time thermal cycler for rapid thermal transfer and optical sensitivity and allows reduction in reagent usage
    Certifications/Compliance RNase, DNase and DNA free
    Type PCR Reaction Tube
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit