Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cepheid™ SmartCycler™ Reaction Tubes

For use with the SmartCycler Real-Time Thermal Cycler

Manufacturer:  Cepheid 9000087

 View more versions of this product

Catalog No. 9000087


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Reaction Tube Rack
Polypropylene
Rack
RNase, DNase and DNA free
8 Pack
SmartCycler™ real-time thermal cycler for rapid thermal transfer and optical sensitivity and allows reduction in reagent usage
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit