Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceranib 1, Tocris Bioscience™

Catalog No. p-4816826
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Ceramidase inhibitor; antiproliferative

Ceramidase inhibitor; inhibits proliferation in SKOV3 ovarian carcinoma cells (IC50 = 3.9 μM). Induces accumulation of ceramide species and decreases sphingosine and sphingosine-1-phosphate (S1P) levels in SKOV3 cells.

Chemical Identifiers

CAS 328076-61-5
Molecular Formula C26H21NO3
Molecular Weight (g/mol) 395.458
InChI Key OJMCCNQNCSCAJV-RVDMUPIBSA-N
Synonym ceranib 1, 3-3-4-methoxy-phenyl-acryloyl-6-methyl-4-phenyl-1h-quinolin-2-one, 3-2e-3-4-methoxyphenyl-1-oxo-2-propen-1-yl-6-methyl-4-phenyl-2 1h-quinolinone, 3-2e-3-4-methoxyphenyl prop-2-enoyl-6-methyl-4-phenyl-1h-quinolin-2-one, 3-e-3-4-methoxyphenyl prop-2-enoyl-6-methyl-4-phenyl-1h-quinolin-2-one
PubChem CID 5761166
IUPAC Name 3-[(E)-3-(4-methoxyphenyl)prop-2-enoyl]-6-methyl-4-phenyl-1H-quinolin-2-one
SMILES CC1=CC2=C(C=C1)NC(=O)C(=C2C3=CC=CC=C3)C(=O)C=CC4=CC=C(C=C4)OC

Specifications

Quantity 10 mg
Formula Weight 395.45
Percent Purity >98%
Chemical Name or Material Ceranib 1
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.