Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cercosporamide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
500μg
This item is not returnable. View return policy
131436-22-1
C16H13NO7
331.28
GEWLYFZWVLXQME-MRXNPFEDSA-N
cercosporamide,--cercosporamide,1s-12-acetyl-3,5,11-trihydroxy-1-methyl-13-oxo-8-oxatricyclo 7.4.0.0 2 ,? trideca-2,4,6,9,11-pentaene-6-carboxamide,?-cercosporamide,d0l1ya,9as-8-acetyl-1,3,7-trihydroxy-9a-methyl-9-oxodibenzofuran-4-carboxamide,9as-8-acetyl-9,9a-dihydro-1,3,7-trihydroxy-9a-methyl-9-oxo-4-dibenzofurancaboxamide,4-dibenzofurancarboxamide, 8-acetyl-9,9a-dihydro-1,3,7-trihydroxy-9a-methyl-9-oxo-, s,9as-8-acetyl-1,3,7-trihydroxy-9a-methyl-9-oxo-9,9a-dihydrodibenzo b,d furan-4-carboxamide
131379
CHEBI:78696
(9aS)-8-acetyl-1,3,7-trihydroxy-9a-methyl-9-oxodibenzofuran-4-carboxamide
CC(=O)C1=C(C=C2C(C1=O)(C3=C(C=C(C(=C3O2)C(=O)N)O)O)C)O
This item is not returnable. View return policy
131436-22-1
C16H13NO7
331.28
GEWLYFZWVLXQME-MRXNPFEDSA-N
cercosporamide,--cercosporamide,1s-12-acetyl-3,5,11-trihydroxy-1-methyl-13-oxo-8-oxatricyclo 7.4.0.0 2 ,? trideca-2,4,6,9,11-pentaene-6-carboxamide,?-cercosporamide,d0l1ya,9as-8-acetyl-1,3,7-trihydroxy-9a-methyl-9-oxodibenzofuran-4-carboxamide,9as-8-acetyl-9,9a-dihydro-1,3,7-trihydroxy-9a-methyl-9-oxo-4-dibenzofurancaboxamide,4-dibenzofurancarboxamide, 8-acetyl-9,9a-dihydro-1,3,7-trihydroxy-9a-methyl-9-oxo-, s,9as-8-acetyl-1,3,7-trihydroxy-9a-methyl-9-oxo-9,9a-dihydrodibenzo b,d furan-4-carboxamide
131379
CHEBI:78696
(9aS)-8-acetyl-1,3,7-trihydroxy-9a-methyl-9-oxodibenzofuran-4-carboxamide
CC(=O)C1=C(C=C2C(C1=O)(C3=C(C=C(C(=C3O2)C(=O)N)O)O)C)O

Potent Mnk2 inhibitor

Potent inhibitor of MAP-kinase interacting kinase-2 (Mnk2) and JAK3 (IC50 values are 11, 31 and 116 nM for Mnk2, JAK3 and Mnk1 respectively). Blocks eIF4E phosphorylation; exhibits antiproliferative and proapoptotic activity in cancer cells in vitro. Also inhibits Pkc1. Orally bioavailable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 500μg
Formula Weight 331.28
Percent Purity >96%
Chemical Name or Material Cercosporamide

Recommended Storage : Store at -20°C