Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceresin Wax, Spectrum™ Chemical
SDP

Catalog Number 18601539
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8001-75-0
C30H50O2
442.73
DSEKYWAQQVUQTP-UHFFFAOYNA-N
2-hydroxy-4,4a,6b,8a,11,11,12b,14a-octamethyl-docosahydropicen-3-one
CC1C(=O)C(O)CC2C1(C)CCC1C2(C)CCC2(C)C3CC(C)(C)CCC3(C)CCC12C
8001-75-0
C30H50O2
442.73
DSEKYWAQQVUQTP-UHFFFAOYNA-N
2-hydroxy-4,4a,6b,8a,11,11,12b,14a-octamethyl-docosahydropicen-3-one
CC1C(=O)C(O)CC2C1(C)CCC1C2(C)CCC2(C)C3CC(C)(C)CCC3(C)CCC12C

CE116, 8001-75-0

Spectrum™ Chemical Ceresin Wax , also known as ceresin or cerin, is a wax that comes from ozokerite through a purifying process. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 100%
Packaging Poly Pail
Quantity 12 kg
Grade Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit