Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Sulfate, Reagent, 85%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
13590-82-4
CeO8S2
332.23
VZDYWEUILIUIDF-UHFFFAOYSA-J
λ⁴-cerium(4+) disulfate
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
13590-82-4
CeO8S2
332.23
VZDYWEUILIUIDF-UHFFFAOYSA-J
λ⁴-cerium(4+) disulfate
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

C1204, 13590-82-4, Ce(SO4)2

Spectrum™ Ceric Sulfate, Reagent is an inorganic compound that is a yellow solid that is water soluble and used as a potent oxidizer. It is used for redox titration in analytical chemistry. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 100%
Insoluble Matter 0.1%
Percent Purity 85%
Packaging Poly Pail
Quantity 2.5 kg