Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 10-Oxomorphine Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco IMPM005061ML
Description
A certified solution standard of the decomposition product measured in manufactured morphine sulfate drug products or APIs using LC or LC/MS.Specifications
67-56-1, 68254-48-8 | |
MFCD09841138 | |
100 μg/mL in methanol | |
299.32 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C17H17NO4 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
10-Ketomorphine | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation