Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 16alpha-Hydroxyestrone, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco H1261ML
Description
A Certified Spiking Solution™ suitable for use as starting material in calibrators, controls, or linearity standards for mass spectrometry-based testing applications in clinical diagnostic testing or endocrinology. 16α-Hydroxyestrone is a major urinary metabolite of estrone and estradiol. This estrogen metabolite is used as an indicator for increased risk of breast cancer along with 2-Hydroxyestrone by ratio comparison of the two analyte levels.Specifications
67-56-1 | |
MFCD00870436 | |
1,3,5(10)-Estratriene-3,16alpha-diol-17-one, 16alpha-3,16-Dihydroxyestra-1,3,5(10)-trien-17-one | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C18H22O3 | |
100 μg/mL in methanol | |
286.37 | |
1 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation