Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 2,4-Dinitrotoluene Solution, 1000 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco ERD033S1.2ML
Description
2,4-Dinitrotoluene is grouped under the family of nitroaromatic compounds, which are organic nitro derivatives of analytical research interest. It is regarded as a common environmental contaminant and can cause adverse effects on human health because of its mutagenic and/or carcinogenic nature.Specifications
75-05-8, 121-14-2 | |
MFCD00007172 | |
1000 μg/mL in acetonitrile | |
Ampule | |
1.2 mL |
C7H6N2O4 | |
UN 1648 3 / PGII | |
182.13 | |
-20°C | |
2°C (35.6°F) - closed cup |
Safety and Handling
H332-H319-H350-H412
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation