Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 2,5-Dimethoxy-4-ethylphenylethylamine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco E0831ML
Description
2C-E, or 2,5-Dimethoxy-4-ethylphenethylamine, is a psychedelic drug and phenethylamine of the 2C series. Sold under street names including 2C-E or Eternity, the designer drug produces vivid visual hallucinations similar to those of LSD. This certified solution standard is suitable as a starting material for calibrators or controls in phenethylamine testing methods by GC/MS or LC/MS for applications in clinical toxicology,forensic analysis, or urine drug testing.Specifications
67-56-1, 923013-67-6 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
245.75 | |
-20°C | |
9.7°C (49.5°F) |
C12H19NO2 · HCl | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation