Learn More
CERILLIANT™ 2,5-Dimethoxy-4-isopropylthiophenethylamine Hydrochloride Solution, 1.0 mg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco T0881ML
Catalog No. 01-002-081
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
2C-T-4 is a psychedelic phenethylamine in the 2C family of recreational drugs. Sold under street names such as Blue Mystic and 7th Heaven, 2C-T-4 produces intense hallucinogenic effects that can last 8 to 16 hours after dosing. This certified Snap-N-Spike™ solution standard is suitable for 2C amine testing methods by GC/MS or LC/MS for applications in clinical toxicology, forensic analysis, or urine drug testing.
Specifications
Specifications
67-56-1, 868738-44-7
UN 1230 - class 3 - PG 2 - Methanol
2C-T-4 hydrochloride solution
Ampule
1 mL
C13H21NO2S · HCl
1.0 mg/mL in methanol (as free base)
291.84
-20°C
9°C (48.2°F)
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation