Learn More
CERILLIANT™ 2,5-Dimethoxy-4-methylphenethylamine-13C, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1061ML
Description
A stable-labeled internal standard suitable for quantification of 2C-D levels in GC/MS or LC/MS phenethylamine testing methods for applications in clinical toxicology, urine drug testing, or forensic analysis. 2C-D, or 2,5-Dimethoxy-4-methylphenethylamine, is a psychedelic drug and stimulant. 2C-D is part of the 2C series of phenethylamines and is sold under street names such as 2C-D or 2C-M.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
2C-D-13C,D3 hydrochloride solution | |
Ampule | |
1 mL |
C1013CH14D3NO2 · HCl | |
100 μg/mL in methanol (as free base) | |
235.73 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation