Learn More
CERILLIANT™ 2,5-Dimethoxy-4-nitrophenethylamine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco N0861ML
Catalog No. 01-001-751
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
2C-N, or 2,5-Dimethoxy-4-nitrophenethylamine, is a psychedelic drug and phenethylamine of the 2C series. Sold under the street name 2C-N, this designer drug produces vivid visual hallucinations similar to those of LSD. This certified solution standard is suitable as a starting material for calibrators or controls in phenethylamine testing methods by GC/MS or LC/MS for applications in clinical toxicology, forensic analysis, or urine drug testing.
Specifications
Specifications
67-56-1, 874140-40-6
UN 1230 - class 3 - PG 2 - Methanol
2C-N hydrochloride solution
Ampule
1 mL
C10H14N2O4 · HCl
1.0 mg/mL in methanol (as free base)
262.69
-20°C
9°C (48.2°F)
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation